missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Septin-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Septin-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7552-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029340

  • € 3405.00 / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001008491.1
Septin-2 Protein
Research Use Only
42980
Recombinant Protein
E. coli
MSKQQPTQFI NPETPGYVGF ANLPNQVHRK SVKKGFEFTL MVVGESGLGK STLINSLFLT DLYPERVIPG AAEKIERTVQ IEASTVEIEE RGVKLRLTVV DTPGYGDAIN CRDCFKTIIS YIDEQFERYL HDESGLNRRH IIDNRVHCCF YFISPFGHGL KPLDVAFMKA IHNKVNIVPV IAKADTLTLK ERERLKKRIL DEIEEHNIKI YHLPDAESDE DEDFKEQTRL LKASIPFSVV GSNQLIEAKG KKVRGRLYPW GVVEVENPEH NDFLKLRTML ITHMQDLQEV TQDLHYENFR SERLKRGGRK VENEDMNKDQ ILLEKEAELR RMQEMIARMQ AQMQMQMQGG DGDGGALGHH V
Greater than 90% as determined by SDS-PAGE.
4735
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers