Learn More
enQuireBio™ Recombinant Human Serine / arginine-rich splicing factor 10 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine/arginine-rich splicing factor 10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7486-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001177934.1 | |
Serine / arginine-rich splicing factor 10 Protein | |
Research Use Only | |
SRSF10 | |
Recombinant Protein | |
E. coli | |
MSRYLRPPNT SLFVRNVADD TRSEDLRREF GRYGPIVDVY VPLDFYTRRP RGFAYVQFED VRDAEDALHN LDRKWICGRQ IEIQFAQGDR KTPNQMKAKE GRNVYSSSRY DDYDRYRRSR SRSYERRRSR SRSFDYNYRR SYSPRNSRPT GRPRRSRSHS DNDRPNCSWN TQYSSAYYTS RKI | |
Greater than 90% as determined by SDS-PAGE. |
10772 | |
50 μg | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |