missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Serine / arginine-rich splicing factor 10 Protein

A DNA sequence encoding the Homo sapiens (Human) Serine/arginine-rich splicing factor 10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7486-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16088960

  • € 530.00 / 100µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001177934.1
Serine / arginine-rich splicing factor 10 Protein
Research Use Only
SRSF10
Recombinant Protein
E. coli
MSRYLRPPNT SLFVRNVADD TRSEDLRREF GRYGPIVDVY VPLDFYTRRP RGFAYVQFED VRDAEDALHN LDRKWICGRQ IEIQFAQGDR KTPNQMKAKE GRNVYSSSRY DDYDRYRRSR SRSYERRRSR SRSFDYNYRR SYSPRNSRPT GRPRRSRSHS DNDRPNCSWN TQYSSAYYTS RKI
Greater than 90% as determined by SDS-PAGE.
10772
100 μg
Not determined.
Full Length of Isoform 3
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers