missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SERPINA3 Protein

A cDNA sequence encoding the SERPINA3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13456-HIS-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933629

  • € 120.00 / 5µg

Estimated Shipment: 06-02-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

SERPINA3 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Lyophilized from 0.02M NH4HCO3.
Greater than 99.0%.
Research Use Only
Recombinant Protein
E. coli
MERMLPLLAL GLLAAGFCPA VLCHPNSPLD EENLTQENQD RGTHVDLGLA SANVDFAFSL YKQLVLKAPD KNVIFSPLSI STALAFLSLG AHNTTLTEIL KGLKFNLTET SEAEIHQSFQ HLLRTLNQSS DELQLSMGNA MFVKEQLSLL DRFTEDAKRL YGSEAFATDF QDSAAAKKLI NDYVKNGTRG KITDLIKDLD SQTMMVLVNY IFFKAKWEMP FDPQDTHQSR FYLSKKKWVM VPMMSLHHLT IPYFRDEELS CTVVELKYTG NASALFILPD QDKMEEVEAM LLPETLKRWR DSLEFREIGE LYLPKFSISR DYNLNDILLQ LGIEEAFTSK ADLSGITGAR NLAVSQVVHK AVLDVFEEGT EASAATAVKI TLLSALVETR TIVRFNRPFL MIIVPTDTQN IFFMSKVTNP KQA
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers