missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SERPINB8 Protein

A cDNA sequence encoding the SERPINB8 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13463-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933649

  • € 4090.00 / 1mg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SERPINB8 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMDDLCEA NGTFAISLFK ILGEEDNSRN VFFSPMSISS ALAMVFMGAK GSTAAQMSQA LCLYKDGDIH RGFQSLLSEV NRTGTQYLLR TANRLFGEKT CDFLPDFKEY CQKFYQAELE ELSFAEDTEE CRKHINDWVA EKTEGKISEV LDAGTVDPLT KLVLVNAIYF KGKWNEQFDR KYTRGMLFKT NEEKKTVQMM FKEAKFKMGY ADEVHTQVLE LPYVEEELSM VILLPDDNTD LAVVEKALTY EKFKAWTNSE KLTKSKVQVF LPRLKLEESY DLEPFLRRLG MIDAFDEAKA DFSGMSTEKN VPLSKVAHKC FVEVNEEGTE AAAATAVVRN SRCSRMEPRF CADHPFLFFI RHHKTNCILF CGRFSSP
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SERPINB8 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers