missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SERPINB8 Protein
A cDNA sequence encoding the SERPINB8 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13463-20ug
Additional Details : Weight : 0.01000kg
Specifications
SERPINB8 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDDLCEA NGTFAISLFK ILGEEDNSRN VFFSPMSISS ALAMVFMGAK GSTAAQMSQA LCLYKDGDIH RGFQSLLSEV NRTGTQYLLR TANRLFGEKT CDFLPDFKEY CQKFYQAELE ELSFAEDTEE CRKHINDWVA EKTEGKISEV LDAGTVDPLT KLVLVNAIYF KGKWNEQFDR KYTRGMLFKT NEEKKTVQMM FKEAKFKMGY ADEVHTQVLE LPYVEEELSM VILLPDDNTD LAVVEKALTY EKFKAWTNSE KLTKSKVQVF LPRLKLEESY DLEPFLRRLG MIDAFDEAKA DFSGMSTEKN VPLSKVAHKC FVEVNEEGTE AAAATAVVRN SRCSRMEPRF CADHPFLFFI RHHKTNCILF CGRFSSP | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SERPINB8 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT. |