missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SERPINH1 Protein

A cDNA sequence encoding the SERPINH1 was constructed and used to recombinantly synthesize the protein.

€ 137.00 - € 2725.00

Specifications

Name SERPINH1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15943669
View Documents Promotion Details
enQuireBio™
QP13469-10UG
10 μg
€ 137.00
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15963669
View Documents Promotion Details
enQuireBio™
QP13469-50UG
50 μg
€ 212.00
50µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15953669
View Documents Promotion Details
enQuireBio™
QP13469-1MG
1 mg
€ 2725.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

SERPINH1 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The SERPINH1 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAAEVKKPAA AAAPGTAEKL SPKAATLAER SAGLAFSLYQ AMAKDQAVEN ILVSPVVVAS SLGLVSLGGK ATTASQAKAV LSAEQLRDEE VHAGLGELLR SLSNSTARNV TWKLGSRLYG PSSVSFADDF VRSSKQHYNC EHSKINFRDK RSALQSINEW AAQTTDGKLP EVTKDVERTD GALLVNAMFF KPHWDEKFHH KMVDNRGFMV TRSYTVGVMM MHRTGLYNYY DDEKEKLQIV EMPLAHKLSS LIILMPHHVE PLERLEKLLT KEQLKIWMGK MQKKAVAISL PKGVVEVTHD LQKHLAGLGL TEAIDKNKAD LSRMSGKKDL YLASVFHATA FELDTDGNPF DQDIYGREEL RSPKLFYADH PFIFLVRDTQ SGSLLFIGRL VRPKGDKMRD EL
Greater than 90.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications