missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Sex comb on midleg-like protein 1

A DNA sequence encoding the Homo sapiens (Human) Sex comb on midleg-like protein 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7903-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031501

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001273337.1
Sex comb on midleg-like protein 1
Research Use Only
SCML4
Recombinant Protein
E. coli
MPCQRTAWIG CDRQRTPPFH WREIKSRVLM TPLALSPPRS TPEPDLSSIP QDAATVPSLA APQALTVCLY INKQANAGPY LERKKVQQLP EHFGPERPSA VLQQAVQACI DCAHQQKLVF SLVKQGYGGE MVSVSASFDG KQHLRSLPVV NSIGYVLRFL AKLCRSLLCD DLFSHQPFPR GCSASEKVQE KEEGRMESVK TVTTEEYLVN PVGMNRYSVD TSASTFNHRG SLHPSSSLYC KRQNSGDSHL GGGPAATAGG PRTSPMSSGG PSAPGLRPPA SSPKRNTTSL EGNRCGNVMH ASASH
Greater than 90% as determined by SDS-PAGE.
256380
500 μg
Not determined.
Full Length of Isoform 3
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers