missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SFRP2 Protein

A cDNA sequence encoding the SFRP2 was constructed and used to recombinantly synthesize the protein.

€ 137.00 - € 4090.00

Specifications

Name SFRP2 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15963699
View Documents Promotion Details
enQuireBio™
QP13477-5UG
5 μg
€ 137.00
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15953699
View Documents Promotion Details
enQuireBio™
QP13477-20UG
20 μg
€ 220.00
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15943699
View Documents Promotion Details
enQuireBio™
QP13477-1MG
1 mg
€ 4090.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

SFRP2 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SFRP2 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSARGLFLF GQPDFSYKRS NCKPIPVNLQ LCHGIEYQNM RLPNLLGHET MKEVLEQAGA WIPLVMKQCH PDTKKFLCSL FAPVCLDDLD ETIQPCHSLC VQVKDRCAPV MSAFGFPWPD MLECDRFPQD NDLCIPLASS DHLLPATEEA PKVCEACKNK NDDDNDIMET LCKNDFALKI KVKEITYINR DTKIILETKS KTIYKLNGVS ERDLKKSVLW LKDSLQCTCE EMNDINAPYL VMGQKQGGEL VITSVKRWQK GQREFKRISR SIRKLQC
Greater than 85.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications