missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SHBG Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | SHBG Protein |
| Quantity | 10 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | Human |
| Protein Tag | His |
| Sequence | AAQPARRARR TKLLLLLLLL LRHTRQGWAL RPVLPTQSAH DPPAVHLSNG PGQEPIAVMT FDLTKITKTS SSFEVRTWDP EGVIFYGDTN PKDDWFMLGL RDGRPEIQLH NHWAQLTVGA GPRLDDGRWH QVEVKMEGDS VLLEVDGEEV LRLRQVSGPL TSKRHPIMRI ALGGLLFPAS NLRLPLVPAL DGCLRRDSWL DKQAEISASA PTSLRSCDVE SNPGIFLPPG TQAEFNLRDI PQPHAEPWAF SLDLGLKQAA GSGHLLALGT PENPSWLSLH LQDQKVVLSS GSGPGLDLPL VLGLPLQLKL SMSRVVLSQG SKMKALALPP LGLAPLLNLW AKPQGRLFLG ALPGEDSSTS FCLNGLWAQG QRLDVDQALN RSHEIWTHSC PQSPGNGTDA SHSRGGPEQKLISEEDLNSA VDHHHHHH |
| Buffer | SHBG filtered solution at a concentration of 55.6microg/ml in certified FBS (Fetal Bovine Serum 5%). |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction