missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Siglec-7/CD328 Fc Chimera Protein

Product Code. 18388367
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18388367

Brand: R&D Systems™ 1138SL050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by the ability of the immobilized protein to support the adhesion of human red blood cells. Kelm, S. et al. (1994) Current Biology 4:965. The ED50 for this effect is 0.5-2 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y286
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl.
Gene ID (Entrez) 27036
Molecular Weight (g/mol) M.W. (Observed): 90-100 kDa, reducing conditions; M.W. (theoretical): 64 kDa (monomer)
Name Siglec-7/CD328 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human Siglec-7/CD328 protein Human Siglec-7 (Gln19-Gly357) (N-terminus) Accession # Q9Y286 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol SIGLEC7
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Siglec-7/CD328
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.