missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human SKA1 His Protein

Product Code. 18131889 Shop All Bio Techne Products
Click to view available options
Quantity:
0.01 mg
0.05 mg
Unit Size:
0.01mg
0.05mg
This item is not returnable. View return policy

Product Code. 18131889

Brand: Novus Biologicals™ NBP2516100.05MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-255 of Human SKA1 The Recombinant Human SKA1 Protein is derived from E. coli. The Recombinant Human SKA1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifica

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0) containing 10% glycerol 0.1M NaCl
Gene ID (Entrez) 220134
Molecular Weight (g/mol) TMW: 31.9kDa
Quantity 0.05 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias C18orf24, chromosome 18 open reading frame 24, MGC10200, spindle and kinetochore associated complex subunit 1, spindle and kinetochore-associated protein 1, spindle and KT (kinetochore) associated 1
Common Name SKA1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >80%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Vedi altri risultati Mostra meno risultati

For research use only and is not approved for use in humans or in clinical diagnosis

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato