Learn More
enQuireBio™ Recombinant Human SNRPC Protein
A cDNA sequence encoding the SNRPC was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 7875.00
Specifications
Name | SNRPC Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15953919
|
enQuireBio™
QP13544-HIS-1UG |
1 μg |
€ 137.00
1µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15963919
|
enQuireBio™
QP13544-HIS-2UG |
2 μg |
€ 137.00
2µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15983919
|
enQuireBio™
QP13544-HIS-5UG |
5 μg |
€ 212.00
5µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933919
|
enQuireBio™
QP13544-HIS-10UG |
10 μg |
€ 212.00
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15973919
|
enQuireBio™
QP13544-HIS-50UG |
50 μg |
€ 1815.00
50µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943919
|
enQuireBio™
QP13544-HIS-1MG |
1 mg |
€ 7875.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SNRPC Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
MPKFYCDYCD TYLTHDSPSV RKTHCSGRKH KENVKDYYQK WMEEQAQSLI DKTTAAFQQG KIPPTPFSAP PPAGAMIPPP PSLPGPPRPG MMPAPHMGGP PMMPMMGPPP PGMMPVGPAP GMRPPMGGHM PMMPGPPMMR PPARPMMVPT RPGMTRPDR | |
Greater than 90% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
His | |
SNRPC is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |