missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Spectrin alpha chain, non-erythrocytic 1 Protein

Product Code. 16075400
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16075400

Brand: enQuireBio™ QP6725ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Spectrin alpha chain, non-erythrocytic 1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001123910.1
Gene ID (Entrez) 6709
Name Spectrin alpha chain, non-erythrocytic 1 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol SPTAN1
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MDPSGVKVLE TAEDIQERRQ QVLDRYHRFK ELSTLRRQKL EDSYRFQFFQ RDAEELEKWI QEKLQIASDE NYKDPTNLQG KLQKHQAFEA EVQANSGAIV KLDETGNLMI SEGHFASETI RTRLMELHRQ WELLLEKMRE KGIKLLQAQK LVQYLRECED VMDWINDKEA IVTSEELGQD LEHVEVLQKK FEEFQTDMAA HEERVNEVNQ FAAKLIQEQH PEEELIKTKQ DEVNAAWQRL KGLALQRQGK LFGAAEVQRF NRDVDETISW IKEKEQLMAS DDFGRDLASV QALLRKHEGL ERDLAALEDK VKALCAEADR LQQSHPLSAT QIQVKREELI TNWEQIRTLA AERHARLNDS YRLQRFLADF RDLTSWVTEM KALINADELA SDVAGAEALL DRHQEHKGEI DAHEDSFKSA DESGQALLAA GHYASDEVRE KLTVLSEERA ALLELWELRR QQYEQCMDLQ LFYRDTEQVD NWMSKQEAFL LNEDLGDSLD SVEALLKKHE DFEKSLSAQE EKITALDEFA TKLIQNNHYA MEDVATRRDA LLSRRNALHE RAMRRRAQLA DSFHLQQFFR
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.