missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Stanniocalcin 2/STC-2 Protein

Product Code. 18757978
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18757978 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18757978 Supplier R&D Systems™ Supplier No. 9405SO050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Stanniocalcin 2/STC-2 Protein is derived from NS0. The Recombinant Human Stanniocalcin 2/STC-2 Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number O76061
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μ filtered solution in PBS
Gene ID (Entrez) 8614
Molecular Weight (g/mol) 31 kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human Stanniocalcin 2/STC-2 protein Thr25-Arg302,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Gene Alias stanniocalcin 2, stanniocalcin-2, STC2, STC-2
Biological Activity 60 to 360ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.