missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STC 2 Protein

A cDNA sequence encoding the STC 2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13613-HIS-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15914149

  • € 4090.00 / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

8614
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
TDATNPPEGP QDRSSQQKGR LSLQNTAEIQ HCLVNAGDVG CGVFECFENN SCEIRGLHGI CMTFLHNAGKFDAQGKSFIK DALKCKAHAL RHRFGCISRK CPAIREMVSQ LQRECYLKHD LCAAAQENTR VIVEMIHFKDLLLHEPYVDL VNLLLTCGEE VKEAITHSVQ VQCEQNWGSL CSILSFCTSA IQKPPTAPPE RQPQVDRTKLSRAHHGEAGH HLPEPSSRET GRGAKGERGS KSHPNAHARG RVGGLGAQGP SGSSEWEDEQ SEYSDIRRAAADYKDDDDK
Greater than 90.0% as determined by SDS-PAGE.
STC 2 Protein
Research Use Only
STC2
Human
His
Filtered (0.4microm) and lyophilized in 0.5 mg/ml in 20mM Tris buffer, 20mM NaCl, pH 7.5.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers