missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STMN1 Protein

A cDNA sequence encoding the STMN1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13620-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934169

  • € 212.00 / 25µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

STMN1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MASSDIQVKE LEKRASGQAF ELILSPRSKE SVPEFPLSPP KKKDLSLEEI QKKLEAAEER RKSHEAEVLK QLAEKREHEK EVLQKAIEEN NNFSKMAEEK LTHKMEANKE NREAQMAAKL ERLREKDKHI EEVRKNKESK DPADETEAD
Greater than 95.0% as determined by SDS-PAGE.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The STMN1 protein solution contains 20mM Tris-HCl, pH 8 and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers