missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SUMO2 Protein

A cDNA sequence encoding the SUMO2 was constructed and used to recombinantly synthesize the protein.

€ 276.00 - € 414.00

Specifications

Gene ID (Entrez) 6613
Name SUMO2 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol SUMO2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15934269
View Documents Promotion Details
enQuireBio™
QP13649-GST-2UG
2 μg
€ 276.00
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15944269
View Documents Promotion Details
enQuireBio™
QP13649-GST-5UG
5 μg
€ 414.00
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

6613
Research Use Only
SUMO2
Human
GST
SUMO2 at 100microg/ml in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol.
SUMO2 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MADEKPKEGV KTENNDHINL KVAGQDGSVV QFKIKRHTPL SKLMKAYCER QGLSMRQIRF RFDGQPINET DTPAQLEMED EDTIDVFQQQ TGGVY
Videos
SDS
Documents

Documents

Product Certifications