missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Syntaxin-11 Protein

Product Code. 16055510
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16055510 1 mg 1mg
16045510 10 μg 10µg
16085510 50 μg 50µg
16035510 100 μg 100µg
16065510 200 μg 200µg
16075510 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16055510 Supplier enQuireBio™ Supplier No. QP6742ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Syntaxin-11, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_003755.2
Gene ID (Entrez) 8676
Name Syntaxin-11 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol STX11
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence MKDRLAELLD LSKQYDQQFP DGDDEFDSPH EDIVFETDHI LESLYRDIRD IQDENQLLVA DVKRLGKQNA RFLTSMRRLS SIKRDTNSIA KAIKARGEVI HCKLRAMKEL SEAAEAQHGP HSAVARISRA QYNALTLTFQ RAMHDYNQAE MKQRDNCKIR IQRQLEIMGK EVSGDQIEDM FEQGKWDVFS ENLLADVKGA RAALNEIESR HRELLRLESR IRDVHELFLQ MAVLVEKQAD TLNVIELNVQ KTVDYTGQAK AQVRKAVQYE EKNPCRTLCC FCCPCLK
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.