missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SYT5 Protein
A cDNA sequence encoding the SYT5 was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 7875.00
Specifications
Name | SYT5 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934309
|
enQuireBio™
QP13662-2UG |
2 μg |
€ 137.00
2µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914309
|
enQuireBio™
QP13662-10UG |
10 μg |
€ 212.00
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924309
|
enQuireBio™
QP13662-1MG |
1 mg |
€ 7875.00
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SYT5 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SYT5 protein solution (0.25 mg/ml) containing 20mM Tris-HCl(pH 8.0) containing 40% glycerol, 0.2M NaCl and 2mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSLGRLQYS LDYDFQSGQL LVGILQAMGL AALDLGGSSD PYVRVYLLPD KRRRYETKVH RQTLNPHFGE TFAFKVPYVE LGGRVLVMAV YDFDRFSRND AIGEVRVPMS SVDLGRPVQA WRELQAAPRE EQEKLGDICF SLRYVPTAGK LTVIVLEAKN LKKMDVGGLS DPYVKVHLLQ GGKKVRKKKT TIKKNTLNPY YNEAFSFEVP CDQVQKVQVE LTVLDYDKLG KNEAIGRVAV GAAAGGAGLR HWADMLANPR RPIAQWHSLR PPDRVRLLPA P | |
Greater than 90.0% as determined by SDS-PAGE. |