missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TAF11 Protein

A DNA sequence encoding the Homo sapiens (Human) TAF11, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7620-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16069840

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001257417.1
TAF11 Protein
Research Use Only
TAF11
Recombinant Protein
E. coli
MDDAHESPSD KGGETGESDE TAAVPGDPGA TDTDGIPEET DGDADVDLKE AAAEEGELES QDVSDLTTVE REDSSLLNPA AKKLKIDTKE KKEKKQKVDE DEIQKMQILV SSFSEEQLNR YEMYRRSAFP KAAIKRLIQS ITGTSVSQNV VIAMSGISKV FVGEVVEEAL DVCEKWGEMP PLQPKHMREA VRRLKSKGQI PNSKHKKIIF F
Greater than 80% as determined by SDS-PAGE.
6882
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers