missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TAF11 Protein
A DNA sequence encoding the Homo sapiens (Human) TAF11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7620-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001257417.1 | |
TAF11 Protein | |
Research Use Only | |
TAF11 | |
Recombinant Protein | |
E. coli | |
MDDAHESPSD KGGETGESDE TAAVPGDPGA TDTDGIPEET DGDADVDLKE AAAEEGELES QDVSDLTTVE REDSSLLNPA AKKLKIDTKE KKEKKQKVDE DEIQKMQILV SSFSEEQLNR YEMYRRSAFP KAAIKRLIQS ITGTSVSQNV VIAMSGISKV FVGEVVEEAL DVCEKWGEMP PLQPKHMREA VRRLKSKGQI PNSKHKKIIF F | |
Greater than 80% as determined by SDS-PAGE. |
6882 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |