missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human TFF2 Protein

Product Code. 18757777
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18757777

Brand: R&D Systems™ 8290TF050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human TFF2 Protein is derived from CHO. The Recombinant Human TFF2 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q03403
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 7032
Molecular Weight (g/mol) 12kDa
Name TFF2
Preparation Method Reconstitute at 500μg/mL in PBS.
Quality Control Testing SDS-PAGE: 17-25kDa, reducing conditions
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human TFF2 protein Glu24-Tyr129
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Glu24
Without Additives Carrier Protein
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.