missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human THAP7 Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | THAP7 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMPRHCSA AGCCTRDTRE TRNRGISFHR LPKKDNPRRG LWLANCQRLD PSGQGLWDPA SEYIYFCSKH FEEDCFELVG ISGYHRLKEG AVPTIFESFS KLRRTTKTKG HSYPPGPAEV SRLRRCRKRC SEGRGPTTPF SPPPPADVTC FPVEEASAPA TLPASPAGRL EPGLSSPFSD LLGPLGAQAD EAGCSAQPSP ERQPSPLEPR PVSPSAYMLR LPPPAGAYIQ NEHSYQVGSA LLWKRRAEAA LDALDKAQRQ LQACKRREQR LRLRLTKLQQ ERAREKRAQA DARQTLKEHV QDFAMQLSSS MA |
| Buffer | THAP7 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction