missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thrombin Protein

Product Code. 15975448
Click to view available options
Quantity:
1 mg
5 mg
500 μg
Unit Size:
1mg
500µg
5mg
This item is not returnable. View return policy

Product Code. 15975448

Brand: enQuireBio™ QP108801mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Thrombin was constructed and used to recombinantly synthesize the protein.

Specifications

Name Thrombin Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity The specific activity was found to be 3005 NIH Units/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species Human
Protein Tag Untagged
Buffer The Factor-IIa contains 50mM sodium Citrate, 0.2M NaCl and 0.1% PEG-8000 pH 6.5.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.