Learn More
enQuireBio™ Recombinant Human Thrombopoietin / THPO Protein
Recombinant Protein
Brand:  enQuireBio™ QP5227-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15945739
  • € 6210.00 / 1mg
Estimated Shipment: 01-07-2024
to see stock.
Specifications
NP_000451.1
Thrombopoietin / THPO Protein
Research Use Only
THPO
Recombinant Protein
E. coli
MSPAPPACDL RVLSKLLRDS HVLHSRLSQC PEVHPLPTPV LLPAVDFSLG EWKTQMEETK AQDILGAVTL LLEGVMAARG QLGPTCLSSL LGQLSGQVRL LLGALQSLLG TQLPPQGRTT AHKDPNAIFL SFQHLLRGKV RFLMLVGGST LCVRRAPPTT AVPSRTSLVL TLNEL
Greater than 90 as determined by SDS-PAGE.
7066
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 5 ng/mL in the MO7e cell proliferaiton assay. The bioactivity acceptance criteria is set at 2 x 10^5 units/mg.
Human
Untagged
10 mM sodium phosphate, pH 7.5