missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Thymidine Kinase 1 Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its ability to phosphorylate thymidine. The specific activity is >550 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | P04183 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, Glycerol, Brij-30 and DTT. |
| Gene ID (Entrez) | 7083 |
| Molecular Weight (g/mol) | M.W. (Observed): 25-30 kDa, reducing conditions; M.W. (theoretical): 26 kDa |
| Name | Thymidine Kinase 1 |
| Quantity | 50 μg |
| Source | E. coli-derived human Thymidine Kinase 1 protein Ser2-Asn234, with C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.1 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction