missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIAL1 Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | TIAL1 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMMEDDGQ PRTLYVGNLS RDVTEVLILQ LFSQIGPCKS CKMITEHTSN DPYCFVEFYE HRDAAAALAA MNGRKILGKE VKVNWATTPS SQKKDTSNHF HVFVGDLSPE ITTEDIKSAF APFGKISDAR VVKDMATGKS KGYGFVSFYN KLDAENAIVH MGGQWLGGRQ IRTNWATRKP PAPKSTQENN TKQLRFEDVV NQSSPKNCTV YCGGIASGLT DQLMRQTFSP FGQIMEIRVF PEKGYSFVRF STHESAAHAI VSVNGTTIEG HVVKCYWGKE SPDMTKNFQQ VDYSQWGQWS QVYGNPQQYG QYMANGWQVP PYGVYGQPWN QQGFGVDQSP SAAWMGGFGA QPPQGQAPPP VIPPPNQAGY GMASYQTQ |
| Buffer | TIAL1 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 50% glycerol, 2mM DTT and 1mM EDTA . |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction