missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TIMP1 Protein

A cDNA sequence encoding the TIMP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13753-HIS-EC-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934589

  • € 212.00 / 10µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TIMP1 Protein
Research Use Only
Recombinant Protein
E. coli
MAPFEPLASG ILLLLWLIAP SRACTCVPPH PQTAFCNSDL VIRAKFVGTP EVNQTTLYQR YEIKMTKMYK GFQALGDAAD IRFVYTPAME SVCGYFHRSH NRSEEFLIAG KLQDGLLHIT TCSFVAPWNS LSLAQRRGFT KTYTVGCEEC TVFPCLSIPC KLQSGTHCLW TDQLLQGSEK GFQSRHLACL PREPGLCTWQ SLRSQIA
Greater than 95.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
TIMP1 protein is supplied in 25mM Sodium acetate pH4.8 and 50% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers