Learn More
enQuireBio™ Recombinant Human TIR domain-containing adapter molecule 2 Protein
Product Code. 16070851
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 16070851
Brand: enQuireBio™ QP7797ec100ug
A DNA sequence encoding the Homo sapiens (Human) TIR domain-containing adapter molecule 2, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_001157940.1
Gene ID (Entrez) 100302736
Name TIR domain-containing adapter molecule 2 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol TICAM2
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence GIGKSKINSC PLSLSWGKRH SVDTSPGYHE SDSKKSEDLS LCNVAEHSNT TEGPTGKQEG AQSVEEMFEE EAEEEVFLKF VILHAEDDTD EALRVQNLLQ DDFGIKPGII FAEMPCGRQH LQNLDDAVNG SAWTILLLTE NFLRDTWCNF QFYTSLMNSV NRQHKYNSVI PMRPLNNPLP RERTPFALQT INALEEESRG FPTQVERIFQ ESVYKTQQTI WKETRNMVQR QFIA
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.