missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human TNF RI/TNFRSF1A Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
TNF RI/TNFRSF1A protein is fully biologically active when compared to standard. The ED50 as determined by its ability to inhibit the TNF-alpha mediated cytotoxicity in the L-929 cells is less than 0.05 μg/mL, corresponding to a specific activity of > 2 x 10^4 IU/mg in the presence of 0.25 ng/mL of rHuTNF-alpha.
Specifications
Specifications
| Accession Number | P19438, P19438, P19438 |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 7132 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 21.2 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 0.1 EU/μg of TNF RI/TNFRSF1A as determined by LAL method. |
| Gene Alias | CD120a, CD120a antigen, FPF, p55, p55-R, p60, sTNF RI, sTNF RI/TNFRSF1A, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1 A isoform beta, tumor necrosis factor receptor superfamily member 1 A, tumor necrosis factor receptor superfamily, member 1 A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor |
| Gene Symbol | TNFRSF1A |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction