Learn More
enQuireBio™ Recombinant Human TNFSF10 / TRAIL / APO-2L Protein
Recombinant Protein
Brand:  enQuireBio™ QP5244-500ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15935769
  • € 1160.00 / 500µg
Estimated Shipment: 28-05-2024
to see stock.
Specifications
NP_001177871.1
TNFSF10 / TRAIL / APO-2L Protein
Research Use Only
TNFSF10
Recombinant Protein
E. coli
MRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVG
Greater than 90 as determined by SDS-PAGE.
8743
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Human TNFSF10 / TRAIL / APO-2L induces RPMI-8226 cell proliferation (typical ED50 is ? 15 ng/ml).
Human
Untagged
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers