missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human TPT1/TCTP His Protein

Product Code. 18513122 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
This item is not returnable. View return policy

Product Code. 18513122

Brand: Novus Biologicals™ NBP1370850.5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-172 of Human TPT1/TCTP The Recombinant Human TPT1/TCTP Protein is derived from E. coli. The Recombinant Human TPT1/TCTP Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13693
For Use With (Application) ELISA
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.1 M NaCl
Gene ID (Entrez) 7178
Molecular Weight (g/mol) 23.9kDa
Name TPT1/TCTP Protein
Purification Method Protein
Quantity 0.5 mg
Source E. Coli
Immunogen TCTP, 1-172aa, Human, His tag, E.coli (NP_003286).
Storage Requirements -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.