missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcription initiation factor IIB Protein
A DNA sequence encoding the Homo sapiens (Human) Transcription initiation factor IIB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6130-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001505.1 | |
Transcription initiation factor IIB Protein | |
Research Use Only | |
GTF2B | |
Recombinant Protein | |
E. coli | |
MASTSRLDAL PRVTCPNHPD AILVEDYRAG DMICPECGLV VGDRVIDVGS EWRTFSNDKA TKDPSRVGDS QNPLLSDGDL STMIGKGTGA ASFDEFGNSK YQNRRTMSSS DRAMMNAFKE ITTMADRINL PRNIVDRTNN LFKQVYEQKS LKGRANDAIA SACLYIACRQ EGVPRTFKEI CAVSRISKKE IGRCFKLILK ALETSVDLIT TGDFMSRFCS NLCLPKQVQM AATHIARKAV ELDLVPGRSP ISVAAAAIYM ASQASAEKRT QKEIGDIAGV ADVTIRQSYR LIYPRAPDLF PTDFKFDTPV DKLPQL | |
Greater than 90% as determined by SDS-PAGE. |
2959 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |