missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transmembrane protein 65 Protein
A DNA sequence encoding the Homo sapiens(Human) Transmembrane protein 65, was expressed in the hosts and tags indicated.
€ 469.00 - € 1705.00
Specifications
Accession Number | NP_919267.2 |
---|---|
Gene ID (Entrez) | 157378 |
Name | Transmembrane protein 65 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16029151
|
enQuireBio™
QP9989-IV-100UG |
100 μg |
€ 1705.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16049151
|
enQuireBio™
QP9989-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16039151
|
enQuireBio™
QP9989-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_919267.2 | |
Transmembrane protein 65 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
157378 | |
Research Use Only | |
TMEM65 | |
Recombinant Protein | |
E. coli | |
MEALNTAQGA RDFIYSLHST ERSCLLKELH RFESIAIAQE KLEAPPPTPG QLRYVFIHNA IPFIGFGFLD NAIMIVAGTH IEMSIGIILG ISTMAAAALG NLVSDLAGLG LAGYVEALAS RLGLSIPDLT PKQVDMWQTR LSTHLGKAVG VTIGCILGMF PLIFFGGGEE DEKLETKS | |
Greater than 85% as determined by SDS-PAGE. |