missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tropomodulin-4 Protein
A DNA sequence encoding the Homo sapiens (Human) Tropomodulin-4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7523-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_037485.2 | |
Tropomodulin-4 Protein | |
Research Use Only | |
TMOD4 | |
Recombinant Protein | |
E. coli | |
MSSYQKELEK YRDIDEDEIL RTLSPEELEQ LDCELQEMDP ENMLLPAGLR QRDQTKKSPT GPLDREALLQ YLEQQALEVK ERDDLVPFTG EKKGKPYIQP KREIPAEEQI TLEPELEEAL AHATDAEMCD IAAILDMYTL MSNKQYYDAL CSGEICNTEG ISSVVQPDKY KPVPDEPPNP TNIEEILKRV RSNDKELEEV NLNNIQDIPI PMLSELCEAM KANTYVRSFS LVATRSGDPI ANAVADMLRE NRSLQSLNIE SNFISSTGLM AVLKAVRENA TLTELRVDNQ RQWPGDAVEM EMATVLEQCP SIVRFGYHFT QQGPRARAAQ AMTRNNELRR QQKKR | |
Greater than 80% as determined by SDS-PAGE. |
29765 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |