missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Trypsin-1 Protein

A cDNA sequence encoding the Trypsin-1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10898-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935508

  • € 137.00 / 5µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Trypsin-1 Protein
Research Use Only
1 unit hydrolyzes 1 micromole of Na-Benzoyl-D,L-arginine 4-nitroanilide (BANI) per minute at 37C, pH 8.0, in the presence of 10mM calcium. The specific activity is >1,000 Units per mg protein as measured under the described conditions.
Human
Untagged
The Trypsin-1 filtered 0.22um solution contains 20mM TRIS-HCl buffer pH 7.2.
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Human
IVGGYNCEEN SVPYQVSLNS GYHFCGGSLI NEQWVVSAGH CYKSRIQVRL GEHNIEVLEG NEQFINAAKI IRHPQYDRKT LNNDIMLIKL SSRAVINARV STISLPTAPP ATGTKCLISG WGNTASSGAD YPDELQCLDA PVLSQAKCEA SYPGKITSNM FCVGFLEGGK DSCQGDSGGP VVCNGQLQGV VSWGDGCAQK NKPGVYTKVY NYVKWIKNTI AANS
Greater than 95% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers