missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TSG101 Protein

A cDNA sequence encoding the TSG101 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13826-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934829

  • € 137.00 / 2µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TSG101 Protein
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMAVS ESQLKKMVSK YKYRDLTVRE TVNVITLYKD LKPVLDSYVF NDGSSRELMN LTGTIPVPYR GNTYNIPICL WLLDTYPYNP PICFVKPTSS MTIKTGKHVD ANGKIYLPYL HEWKHPQSDL LGLIQVMIVV FGDEPPVFSR P
Greater than 90% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
TSG101 protein solution (0.5 mg/ml) containing 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers