missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TXNRD1 / TRXR1 Protein
A DNA sequence encoding the Homo sapiens (Human) TXNRD1 / TRXR1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7539-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001087240.1 | |
TXNRD1 / TRXR1 Protein | |
Research Use Only | |
TXNRD1 | |
Recombinant Protein | |
E. coli | |
MNGPEDLPKS YDYDLIIIGG GSGGLAAAKE AAQYGKKVMV LDFVTPTPLG TRWGLGGTCV NVGCIPKKLM HQAALLGQAL QDSRNYGWKV EETVKHDWDR MIEAVQNHIG SLNWGYRVAL REKKVVYENA YGQFIGPHRI KATNNKGKEK IYSAERFLIA TGERPRYLGI PGDKEYCISS DDLFSLPYCP GKTLVVGASY VALECAGFLA GIGLDVTVMV RSILLRGFDQ DMANKIGEHM EEHGIKFIRQ FVPIKVEQIE AGTPGRLRVV AQSTNSEEII EGEYNTVMLA IGRDACTRKI GLETVGVKIN EKTGKIPVTD EEQTNVPYIY AIGDILEDKV ELTPVAIQAG RLLAQRLYAG STVKCDYENV PTTVFTPLEY GACGLSEEKA VEKFGEENIE VYHSYFWPLE WTIPSRDNNK CYAKIICNTK DNERVVGFHV LGPNAGEVTQ GFAAALKCGL TKKQLDSTIG IHPVCAEVFT TLSVTKRSGA SILQAGC | |
Greater than 90% as determined by SDS-PAGE. |
7296 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 5 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |