missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UDP-N-acetylhexosamine pyrophosphorylase Protein

A DNA sequence encoding the Homo sapiens (Human) UDP-N-acetylhexosamine pyrophosphorylase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7555-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029360

Quantity Price
1 € 1180.00 / 500µg
3 € 1100.47 / 500µg
Estimated Shipment
03-11-2020
Add to basket


Specifications

Specifications

NP_001311042.1
UDP-N-acetylhexosamine pyrophosphorylase Protein
500 μg
Not determined.
Full Length of Isoform AGX1. 
Human
His
Tris-based buffer, 50% glycerol
6675
Greater than 90% as determined by SDS-PAGE.
Research Use Only
UAP1
Recombinant Protein
E. coli
MNINDLKLTL SKAGQEHLLR FWNELEEAQQ VELYAELQAM NFEELNFFFQ KAIEGFNQSS HQKNVDARME PVPREVLGSA TRDQDQLQAW ESEGLFQISQ NKVAVLLLAG GQGTRLGVAY PKGMYDVGLP SRKTLFQIQA ERILKLQQVA EKYYGNKCII PWYIMTSGRT MESTKEFFTK HKYFGLKKEN VIFFQQGMLP AMSFDGKIIL EEKNKVSMAP DGNGGLYRAL AAQNIVEDME QRGIWSIHVY CVDNILVKVA DPRFIGFCIQ KGADCGAKVV EKTNPTEPVG VVCRVDGVYQ VVEYSEISLA TAQKRSSDGR LLFNAGNIAN HFFTVPFLRD VVNVYEPQLQ HHVAQKKIPY VDTQGQLIKP DKPNGIKMEK FVFDIFQFAK KFVVYEVLRE DEFSPLKNAD SQNGKDNPTT ARHALMSLHH CWVLNAGGHF IDENGSRLPA IPRLKDANDV PIQCEISPLI SYAGEGLESY VADKEFHAPL IIDENGVHEL VKNGI
Videos
SDS
Documents