missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UGT8 Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | UGT8 Protein |
| Quantity | 20 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSAKIIIVP PIMFESHMYI FKTLASALHE RGHHTVFLLS EGRDIAPSNH YSLQRYPGIF NSTTSDAFLQ SKMRNIFSGR LTAIELFDIL DHYTKNCDLM VGNHALIQGL KKEKFDLLLV DPNDMCGFVI AHLLGVKYAV FSTGLWYPAE VGAPAPLAYV PEFNSLLTDR MNLLQRMKNT GVYLISRLGV SFLVLPKYER IMQKYNLLPE KSMYDLVHGS SLWMLCTDVA LEFPRPTLPN VVYVGGILTK PASPLPEDLQ RWVNGANEHG FVLVSFGAGV KYLSEDIANK LAGALGRLPQ KVIWRFSGPK PKNLGNNTKL IEWLPQNDLL GHSKIKAFLS HGGLNSIFET MYHGVPVVGI PLFGDHYDTM TRVQAKGMGI LLEWKTVTEK ELYEALVKVI NNPSYRQRAQ KLSEIHKDQP GHPVNRTIYW IDYIIRHNGA HHLRAAVHQI SFCQYFLLDI AFVLLLGAAL LYFLLSWVTK FIYRKIKSLW SRNKHSTVNG HYHNGILNGK YKRNGHIKHE KKVK |
| Buffer | UGT8 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction