missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ULBP4 Protein

A cDNA sequence encoding the ULBP4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13888-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935029

  • € 137.00 / 2µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ULBP4 Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASHSLCFNFTIK SLSRPGQPWC EAQVFLNKNL FLQYNSDNNM VKPLGLLGKK VYATSTWGEL TQTLGEVGRD LRMLLCDIKP QIKTSDPSTL QVEMFCQREA ERCTGASWQF ATNGEKSLLF DAMNMTWTVI NHEASKIKET WKKDRGLEKY FRKLSKGDCD HWLREFLGHW EAMPEPTGRR ST
Greater than 85.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
ULBP4 was filtered (0.4 microm) and lyophilized in 0.05M Acetate buffer pH 4.0, 1% (w/v) Sucrose and 4% (w/v) Mannitol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers