missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Uncharacterized protein C1orf106 Protein
A DNA sequence encoding the Homo sapiens (Human) Uncharacterized protein C1orf106, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9786-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001136041.1 | |
Uncharacterized protein C1orf106 Protein | |
Research Use Only | |
C1orf106 | |
Recombinant Protein | |
Yeast | |
MLQMPKLNEI PPGRAGRREA RGEGRWPGQT GPEAARLEWR AQGQAGGARA PWDSWGSSRL PTQPGPGWSR CPPSLLCALS FQKSTMESKD EVSDTDSGII LQSGPDSPVS PMKELTHAVH KQQRALEARL EACLEELRRL CLREAELTGT LPAEYPLKPG EKAPKVRRRI GAAYKLDDWA LHREDPLSSL ERQLALQLQI TEAARRLCLE ENLSRQARRQ RKHSMLQEEK KLQELQRCLV ERRRNSEPPP AAALPLGREL SASDDSSLSD GLLLEEEESQ VPKPPPESPA PPSRPLPPQT LEGLQPTGPE AGSPERAPVQ NSPWKETSLD HPYEKPRKSS EPWSESSSPA TTPQDGPSAS SLWLLEPASY HVVPIRGVPG QWQGRTSAPA TPEIQGRRGQ SQSLRVDSFR AGPEGRGRSA FPRRRPTHYT VTVPDSCFPA TKPPLPHAAC HSCSEDSGSD VSSISHPTSP GSSSPDISFL QPLSPPKTHR HRGAWVPAGS RELVAHHPKL LLPPGYFPAG RYVVVAESPL PPGEWELRRA APGPAYEEEG TPLRYQRLVP SRSRIVRTPS LKDSPAGR | |
Greater than 90% as determined by SDS-PAGE. |
55765 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |