missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UQCRC2 Protein

A cDNA sequence encoding the UQCRC2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13904-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935079

  • € 7875.00 / 1mg
Estimated Shipment: 02-02-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

UQCRC2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSLKVAPKVK ATAAPAGAPP QPQDLEFTKL PNGLVIASLE NYSPVSRIGL FIKAGSRYED FSNLGTTHLL RLTSSLTTKG ASSFKITRGI EAVGGKLSVT ATRENMAYTV ECLRGDVDIL MEFLLNVTTA PEFRRWEVAD LQPQLKIDKA VAFQNPQTHV IENLHAAAYR NALANPLYCP DYRIGKVTSE ELHYFVQNHF TSARMALIGL GVSHPVLKQV AEQFLNMRGG LGLSGAKANY RGGEIREQNG DSLVHAAFVA ESAVAGSAEA NAFSVLQHVL GAGPHVKRGS NTTSHLHQAV AKATQQPFDV SAFNASYSDS GLFGIYTISQ ATAAGDVIKA AYNQVKTIAQ GNLSNTDVQA AKNKLKAGYL MSVESSECFL EEVGSQALVA GSYMPPSTVL QQIDSVANAD IINAAKKFVS GQKSMAASGN LGHTPFVDEL
Greater than 85% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The UQCRC2 solution contains 20mM Tris-HCl buffer (pH 8.0), 2M Urea, 0.1M NaCl and 10% glycerol.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers