missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Vasohibin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Vasohibin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7744-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055724.1 | |
Vasohibin-1 Protein | |
Research Use Only | |
VASH1 | |
Recombinant Protein | |
E. coli | |
MPGGKKVAGG GSSGATPTSA AATAPSGVRR LETSEGTSAQ RDEEPEEEGE EDLRDGGVPF FVNRGGLPVD EATWERMWKH VAKIHPDGEK VAQRIRGATD LPKIPIPSVP TFQPSTPVPE RLEAVQRYIR ELQYNHTGTQ FFEIKKSRPL TGLMDLAKEM TKEALPIKCL EAVILGMYPS SPEGEGSGLL WASASCSESE GGVG | |
Greater than 90% as determined by SDS-PAGE. |
22846 | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |