missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human VEGF 121 Protein

Product Code. 18603817 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 18603817

Brand: Novus Biologicals™ NBP307103100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human umbilical vein endothelial cells (HUVEC) is between 0.2-0.4 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P15692-9
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in PBS, (pH 7.4)
Gene ID (Entrez) 7422
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 0.1 EU/μg of VEGF 121 AS determined by LAL method.
Gene Alias MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
Gene Symbol VEGFA
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Briefly centrifuge vial prior to opening to bring the contents to the bottom. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to 0.1-1.0 mg/mL Stock solutions should be apportioned into working aliquots and stored (at -20°C.)
Form Purified
Research Category Angiogenesis, Biologically Active Proteins
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein VEGF 121
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.