missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human VIPR2 Fc Chimera Protein

Product Code. 18748126
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18748126

Brand: R&D Systems™ 11003VR050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. When Recombinant Human Growth Hormone-Releasing Factor is immobilized at 2.00 μg/mL, 100 μL/well, Recombinant Human VIPR2 Fc Chimera binds with an ED50 of 7.00-42.0 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P41587.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 7434
Molecular Weight (g/mol) MolecularWeight-observed: 45-60 kDa, under reducing conditions.; MolecularWeight-theroretical: 38 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human VIPR2 protein Recombinant Human VIPR2 (Arg26-Val126) (N-terminus) Accession # P41587.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias FLJ16511, Helodermin-preferring VIP receptor, PACAP type III receptor, PACAPR3, PACAP-R3, PACAP-R-3, Pituitary adenylate cyclase-activating polypeptide type III receptor, vasoactive intestinal peptide receptor 2, vasoactive intestinal polypeptide receptor 2, VIP R2, VIP2R, VIP-R-2, VPAC2, VPCAP2R
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein VIPR2/VPAC2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.