missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human VSIG3 C-Terminal Fc Chimera Protein

Product Code. 18716356
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18716356

Brand: R&D Systems™ 10075VS050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit anti-CD3 antibody induced IL-17 or IFN-gamma secretion by human peripheral blood mononuclear cells (PBMC). The ED50 for this effect is 1-6 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5DX21
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 152404
Molecular Weight (g/mol) M.W. (Observed): 41-51 kDa, reducing conditions; M.W. (theoretical): 37 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human VSIG3 protein Human VSIG3 (Pro144-Gly241) (N-terminus) Accession # Q5DX21 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Specific Reactivity VSIG3/IGSF11
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein VSIG3/IGSF11
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.