missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zinc finger protein 346 Protein

Product Code. 16056680
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16056680

Brand: enQuireBio™ QP6907ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 346, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001295142.1
Gene ID (Entrez) 23567
Name Zinc finger protein 346 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Znf346
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence MEYPAPATVQ AADGGAAGPY SSSELLEGQE PDGVRFDRER ARRLWEAVSG AQPVGREEVE HMIQKNQCLF TNTQCKVCCA LLISESQKLA HYQSKKHANK VKRYLAIHGM ETLKGETKKL DSDQKSSRSK DKNQCCPICN MTFSSPVVAQ SHYLGKTHAK NLKLKQQSTK VEALHQNREM IDPDKFCSLC HATFNDPVMA QQHYVGKKHR KQETKLKLMA RYGRLADPAV TDFPAGKGYP CKTCKIVLNS IEQYQAHVSG FKHKNQSPKT VASSLGQIPM QRQPIQKDST TLED
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.