missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zymogen granule protein 16 homolog B Protein

A DNA sequence encoding the Homo sapiens (Human) Zymogen granule protein 16 homolog B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7869-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031281

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_660295.2
Zymogen granule protein 16 homolog B Protein
Research Use Only
ZG16B
Recombinant Protein
Yeast
GKMYGPGGGK YFSTTEDYDH EITGLRVSVG LLLVKSVQVK LGDSWDVKLG ALGGNTQEVT LQPGEYITKV FVAFQAFLRG MVMYTSKDRY FYFGKLDGQI SSAYPSQEGQ VLVGIYGQYQ LLGIKSIGFE WNYPLEEPTT EPPVNLTYSA NSPVGR
Greater than 90% as determined by SDS-PAGE.
124220
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers